5-Isopropyl-3-(methoxycarbonyl)-2H-cyclohepta[b]furan-2-one ≥98.0%

Supplier: TCI America

I0657-5G
TCI0657-5GEA 162.32 USD
TCI0657-5G
5-Isopropyl-3-(methoxycarbonyl)-2H-cyclohepta[b]furan-2-one ≥98.0%
5-Isopropyl-3-(methoxycarbonyl)-2H-cyclohepta[b]furan-2-one

CAS Number: 99909-62-3
Molecular Formula: C14H14O4
Molecular Weight: 246.26
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 126

Formula: C₁₄H₁₄O₄
Melting Pt: 126 °C
CAS Number: 99909-62-3
https://us.vwr.com/stibo/web/std.lang.all/25/34/28972534.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR