(S,S,S)-Triglycidyl isocyanurate ≥98.0% (by total nitrogen basis)
Supplier: TCI America
I0581-1G
TCI0581-1GEA
152
USD
TCI0581-1G
(S,S,S)-Triglycidyl isocyanurate ≥98.0% (by total nitrogen basis)
(S,S,S)-Triglycidyl isocyanurate
CAS Number: 240408-81-5
MDL Number: MFCD03844799
Molecular Formula: C12H15N3O6
Molecular Weight: 297.27
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 107
Specific rotation [a]20/D: -22 deg (C=0.4, H2O)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: Isocyanuric Acid (S,S,S)-Tris(2,3-epoxypropyl) Ester, Isocyanuric Acid (S,S,S)-Triglycidyl Ester, (S,S,S)-Tris(2,3-epoxypropyl) Isocyanurate
Formula:
C₁₂H₁₅N₃O₆ Melting Pt: 107 °C |
MDL Number:
MFCD03844799 CAS Number: 240408-81-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...