(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥97.0%

Supplier: TCI America

Synonyms: (S)-4-Isopropyl-3-propionyloxazolidin-2-one

I0573-5G I0573-1G
TCI0573-5GEA 157 USD
TCI0573-5G TCI0573-1G
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone ≥97.0%
(S)-(+)-4-Isopropyl-3-propionyl-2-oxazolidinone

CAS Number: 77877-19-1
MDL Number: MFCD00043401
Molecular Formula: C9H15NO3
Molecular Weight: 185.22
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.10
Specific rotation [a]20/D: 89 deg (neat)

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
Density: 1.10 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00043401
CAS Number: 77877-19-1
https://us.vwr.com/stibo/web/std.lang.all/24/51/28972451.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR