5-Chloro-2,1,3-benzothiadiazol-4-ylamine ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 5-Chlorobenzo[c][1,2,5]thiadiazol-4-amine

A1513-5G
TCA1513-005GEA 39 USD
TCA1513-005G
5-Chloro-2,1,3-benzothiadiazol-4-ylamine ≥97.0% (by titrimetric analysis)
5-Chloro-2,1,3-benzothiadiazol-4-ylamine

CAS Number: 30536-19-7
MDL Number: MFCD00465908
Molecular Formula: C6H4ClN3S
Molecular Weight: 185.63
Purity/Analysis Method: 97.0% (T)
Form: Crystal
Melting point (°C): 89


Other Product Names / Synonyms: 4-Amino-5-chloropiazthiole

Formula: C₆H₄ClN₃S
MW: 185.6341 g/mol
Melting Pt: 89 °C
MDL Number: MFCD00465908
CAS Number: 30536-19-7
https://us.vwr.com/stibo/web/std.lang.all/95/76/28929576.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR