(S)-2-Isopropylamino-3-methyl-1-butanol ≥97.0%

Supplier: TCI America

Synonyms: (S)-N-Isopropyl-2-(3-methyl-1-butanol)amine

I0480-5G
TCI0480-005GEA 296 USD
TCI0480-005G
(S)-2-Isopropylamino-3-methyl-1-butanol ≥97.0%
(S)-2-Isopropylamino-3-methyl-1-butanol

CAS Number: 112211-88-8
MDL Number: MFCD00671550
Molecular Formula: C8H19NO
Molecular Weight: 145.25
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.88
Specific rotation [a]20/D: -3 deg (C=1, EtOH)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-N-Isopropyl-2-(3-methyl-1-butanol)amine

Formula: C₈H₁₉NO
MW: 145.25 g/mol
Density: 0.88 (20 °C)
MDL Number: MFCD00671550
CAS Number: 112211-88-8
https://us.vwr.com/stibo/web/std.lang.all/23/37/28972337.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR