TCIAI0475

I0475-25G
TCI0475-025GEA 745.28 USD
TCI0475-025G
TCIAI0475
(stabilized with HQ)
CAS Number: 13816-04-1
MDL Number: MFCD00671547
Molecular Formula: C6H7NS
Molecular Weight: 125.19
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 63
Specific Gravity (20/20): 1.09
Storage Temperature: 0°C
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR