2-Isobutyl-4-methylthiazole ≥96.0%

Supplier: TCI America

I0442-5ML
TCI0442-005MLEA 133 USD
TCI0442-005ML
2-Isobutyl-4-methylthiazole ≥96.0%
2-Isobutyl-4-methylthiazole

CAS Number: 61323-24-8
MDL Number: MFCD00053138
Molecular Formula: C8H13NS
Molecular Weight: 155.26
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Specific Gravity (20/20): 0.98

Formula: C₈H₁₃NS
MW: 155.2605 g/mol
Boiling Pt: 189 °C
Density: 0,98 (20 °C)
MDL Number: MFCD00053138
CAS Number: 61323-24-8

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR