N-(1-Isopropoxyethyl)acetamide ≥90.0%

Supplier: TCI America

I0441-25G
TCI0441-025GEA 41.6 USD
TCI0441-025G
N-(1-Isopropoxyethyl)acetamide ≥90.0%
N-(1-Isopropoxyethyl)acetamide

CAS Number: 115910-75-3
MDL Number: MFCD00191526
Molecular Formula: C7H15NO2
Molecular Weight: 145.20
Purity/Analysis Method: 90.0% (GC)
Form: Clear Liquid
Flash Point (°C): 89

Formula: C₇H₁₅NO₂
Flash Pt: 89 °C
MDL Number: MFCD00191526
CAS Number: 115910-75-3
https://us.vwr.com/stibo/web/std.lang.all/22/89/28972289.jpg

Order Now

Specification Test Results

Purity/Analysis Method 90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR