Triglycidyl isocyanurate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,3,5-Triglycidyl isocyanurate

I0428-500G I0428-100G I0428-25G
TCI0428-025GEA 41.6 USD
TCI0428-025G TCI0428-500G TCI0428-100G
Triglycidyl isocyanurate ≥98.0% (by GC, titration analysis)
Triglycidyl isocyanurate

CAS Number: 2451-62-9
MDL Number: MFCD00080670
Molecular Formula: C12H15N3O6
Molecular Weight: 297.27
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 108
Flash Point (°C): 170


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: Isocyanuric Acid Tris(2,3-epoxypropyl) Ester, Isocyanuric Acid Triglycidyl Ester, Tris(2,3-epoxypropyl) Isocyanurate

Formula: C₁₂H₁₅N₃O₆
MW: 297.2640 g/mol
Melting Pt: 108 °C
Flash Pt: 170 °C
MDL Number: MFCD00080670
CAS Number: 2451-62-9
https://us.vwr.com/stibo/web/std.lang.all/22/70/28972270.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR