2-Isobutyl-3-methylpyrazine ≥98.0%
Supplier: TCI America
Synonyms:
2-Methyl-3-isobutylpyrazine
I0337-5ML
TCI0337-005MLEA
105.51
USD
TCI0337-005ML
2-Isobutyl-3-methylpyrazine ≥98.0%
2-Isobutyl-3-methylpyrazine
CAS Number: 13925-06-9
MDL Number: MFCD00059770
Molecular Formula: C9H14N2
Molecular Weight: 150.23
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 92
Flash Point (°C): 82
Specific Gravity (20/20): 0.94
Other Product Names / Synonyms: 2-Methyl-3-isobutylpyrazine
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...