cis-3-Hexenyl isovalerate ≥98.0%
Supplier: TCI America
Synonyms:
(Z)-Hex-3-en-1-yl 3-methylbutanoate
, Isovaleric Acid cis-3-Hexenyl Ester
I0307-25ML
TCI0307-025MLEA
40
USD
TCI0307-025ML
cis-3-Hexenyl isovalerate ≥98.0%
cis-3-Hexenyl isovalerate
CAS Number: 35154-45-1
MDL Number: MFCD00036533
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 199
Flash Point (°C): 101
Specific Gravity (20/20): 0.88
Other Product Names / Synonyms: Isovaleric Acid cis-3-Hexenyl Ester
Formula:
C₁₁H₂₀O₂ MW: 184.28 g/mol Boiling Pt: 199 °C Density: 0.88 (20 °C) Flash Pt: 101 °C |
MDL Number:
MFCD00036533 CAS Number: 35154-45-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...