Monoethyl itaconate ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: beta-Ethyl Itaconate4-Ethyl MethylenesuccinateItaconic Acid Monoethyl Ester

I0268-25G
TCI0268-025GEA 93.33 USD
TCI0268-025G
Monoethyl itaconate ≥97.0% (by GC, titration analysis)
Monoethyl itaconate

CAS Number: 57718-07-7
MDL Number: MFCD00059138
Molecular Formula: C7H10O4
Molecular Weight: 158.15
Purity/Analysis Method: 97.0% (GC,T)
Form: Crystal
Boiling point (°C): 153
Melting point (°C): 56
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Copolymerization Monomers
Other Product Names / Synonyms: beta-Ethyl Itaconate, 4-Ethyl Methylenesuccinate, Itaconic Acid Monoethyl Ester

Formula: C₇H₁₀O₄
MW: 158.15 g/mol
Boiling Pt: 153 °C
Melting Pt: 56 °C
MDL Number: MFCD00059138
CAS Number: 57718-07-7
https://us.vwr.com/stibo/web/std.lang.all/20/79/28972079.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR