Indigo ≥97.0% (by total nitrogen basis)

Supplier: TCI America
Warning

Synonyms: 2-(1,3-Dihydro-3-oxo-2H-indol-2-ylidene)-1,2-dihydro-3H-indol-3-one Pigment Blue 66 2,2'-Bis(2,3-dihydro-3-oxoindolyliden) Indigotin Vat. Blue 1

I0212-25G
TCI0212-025GEA 66.99 USD
TCI0212-025G
Indigo ≥97.0% (by total nitrogen basis)
Indigo

CAS Number: 482-89-3
MDL Number: MFCD00005722
Molecular Formula: C16H10N2O2
Molecular Weight: 262.27
Purity/Analysis Method: 97.0% (N)
Form: Crystal
Lambda max.: 603 nm (CHCl3)


Product Categories: Bioscience, Stains & Dyes
Other Product Names / Synonyms: Indigotin, Vat Blue 1

Formula: C₁₆H₁₀N₂O₂
MW: 262.27 g/mol
Density: 1.48 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00005722
CAS Number: 482-89-3
EINECS: 207-586-9
Merck Index: 12,04977
https://us.vwr.com/stibo/web/std.lang.all/20/10/28972010.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR