TCIAI0195

I0195-25ML
TCI0195-025MLEA 87.44 USD
TCI0195-025ML
TCIAI0195
CAS Number: 7774-44-9
MDL Number: MFCD00046351
Molecular Formula: C11H20O2
Molecular Weight: 184.28
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 223
Flash Point (°C): 106
Specific Gravity (20/20): 0.93

Other Product Names / Synonyms: Isovaleric Acid Cyclohexyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR