Diisopropyl ether ≥99.0% stabilized
Supplier: TCI America
|
Danger
|
Synonyms:
Isopropyl ether
I0172-500ML
TCI0172-500MLEA
46.39
USD
TCI0172-500ML
Diisopropyl ether ≥99.0% stabilized
Diisopropyl ether
(stabilized with BHT)
CAS Number: 108-20-3
MDL Number: MFCD00008880
Molecular Formula: C6H14O
Molecular Weight: 102.18
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 68
Melting point (°C): -60
Flash Point (°C): -28
Specific Gravity (20/20): 0.72
Other Product Names / Synonyms: Diisopropyl Ether
Formula:
(CH₃)₂CHOCH(CH₃)₂ MW: 102.18 g/mol Boiling Pt: 68 °C Melting Pt: –60 °C Density: 0.72 (20 °C) Flash Pt: –28 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008880 CAS Number: 108-20-3 EINECS: 203-560-6 UN: 1159 ADR: 3,II Merck Index: 13,05232 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...