1-Cyanovinyl acetate ≥97.0% stabilized

Supplier: TCI America

Synonyms: 2-Acetoxyacrylonitrile1-Acetoxy-1-cyanoethyleneAcetic Acid 1-Cyanovinyl Ester

A1477-25G A1477-5G
TCA1477-005GEA 311.49 USD
TCA1477-005G TCA1477-25G
1-Cyanovinyl acetate ≥97.0% stabilized
1-Cyanovinyl acetate

(stabilized with TBC)
CAS Number: 3061-65-2
MDL Number: MFCD00001868
Molecular Formula: C5H5NO2
Molecular Weight: 111.10
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 173
Flash Point (°C): 69
Specific Gravity (20/20): 1.06
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2-Acetoxyacrylonitrile, 1-Acetoxy-1-cyanoethylene, Acetic Acid 1-Cyanovinyl Ester

Formula: C₅H₅NO₂
MW: 111.10 g/mol
Boiling Pt: 173 °C
Density: 1.06 (20 °C)
Flash Pt: 69 °C
MDL Number: MFCD00001868
CAS Number: 3061-65-2
https://us.vwr.com/stibo/web/std.lang.all/94/93/28929493.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR