N-(2-Hydroxyethyl)acrylamide ≥98.0% (by GC, titration analysis) stabilized
Supplier: TCI America
H1262-500G
H1262-25G
TCH1262-25GEA
42.4
USD
TCH1262-25G
TCH1262-500G
N-(2-Hydroxyethyl)acrylamide ≥98.0% (by GC, titration analysis) stabilized
N-(2-Hydroxyethyl)acrylamide
(stabilized with MEHQ)
CAS Number: 7646-67-5
Molecular Formula: C5H9NO2
Molecular Weight: 115.13
Purity/Analysis Method:
Form: Clear Liquid
Boiling point (°C): 130
Specific Gravity (20/20): 1.12
Storage Temperature: 0-10°C
Formula:
C₅H₉NO₂ MW: 115.13 g/mol Boiling Pt: 130 °C Density: 1.12 (20 °C) |
MDL Number:
MFCD11110645 CAS Number: 7646-67-5 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...