17α-Hydroxyprogesterone ≥98.0% (by HPLC)

Synonyms: 17α-Hydroxy-4-pregnene-3,20-dione

H1250-5G H1250-25G
TCH1250-5GEA 85.8 USD
TCH1250-5G TCH1250-25G
17α-Hydroxyprogesterone ≥98.0% (by HPLC)
17α-Hydroxyprogesterone

Form: Crystal
Specific rotation [a]20/D: 90 deg (C=1, CHCl3)
Product Categories: Bioscience, Pregnanes (Progestogen), Steroids

Formula: C₂₁H₃₀O₃
MW: 330.46 g/mol
Melting Pt: 220 °C
MDL Number: MFCD00003659
CAS Number: 68-96-2
https://us.vwr.com/stibo/web/std.lang.all/11/75/28971175.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR