2'-Hydroxy-4'-methoxypropiophenone ≥98.0%
Supplier: TCI America
Synonyms:
1-(2-Hydroxy-4-methoxyphenyl)propan-1-one
H1207-5G
TCH1207-5GEA
80.15
USD
TCH1207-5G
2'-Hydroxy-4'-methoxypropiophenone ≥98.0%
2'-Hydroxy-4'-methoxypropiophenone
CAS Number: 6270-44-6
MDL Number: MFCD04037453
Molecular Formula: C10H12O3
Molecular Weight: 180.20
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 58
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...