(R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane ≥94.0%

Supplier: TCI America

Synonyms: (R)-2-(2,2-Dimethyl-1,3-dioxolan-4-yl)ethanol

H1188-1G
TCH1188-1GEA 608 USD
TCH1188-1G
(R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane ≥94.0%
(R)-4-(2-Hydroxyethyl)-2,2-dimethyl-1,3-dioxolane

CAS Number: 70005-89-9
MDL Number: MFCD02682966
Molecular Formula: C7H14O3
Molecular Weight: 146.19
Purity/Analysis Method: 94.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 106
Specific Gravity (20/20): 1.05


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (R)-2-(2,2-Dimethyl-1,3-dioxolan-4-yl)ethanol

Formula: C₇H₁₄O₃
MW: 146.19 g/mol
Boiling Pt: 110 °C
Density: 1.05 (20 °C)
Flash Pt: 106 °C
MDL Number: MFCD02682966
CAS Number: 70005-89-9
https://us.vwr.com/stibo/web/std.lang.all/11/28/28971128.jpg

Order Now

Specification Test Results

Purity/Analysis Method 94.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR