2-Hydroxyisobutyramide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 2-Hydroxy-2-methylpropionamide2-Hydroxy-2-methylpropanamide

H1181-25G
TCH1181-25GEA 134 USD
TCH1181-25G
2-Hydroxyisobutyramide ≥98.0% (by total nitrogen basis)
2-Hydroxyisobutyramide

CAS Number: 13027-88-8
MDL Number: MFCD08063365
Molecular Formula: C4H9NO2
Molecular Weight: 103.12
Purity/Analysis Method: 98.0% (N)
Form: Crystal
Boiling point (°C): 260
Melting point (°C): 93


Other Product Names / Synonyms: 2-Hydroxy-2-methylpropionamide

Formula: C₄H₉NO₂
MW: 103.12 g/mol
Boiling Pt: 260 °C
Melting Pt: 93 °C
MDL Number: MFCD08063365
CAS Number: 13027-88-8
https://us.vwr.com/stibo/web/std.lang.all/11/24/28971124.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR