TCIAH1065

H1065-25G
TCH1065-25GEA 210.22 USD
TCH1065-25G
TCIAH1065
CAS Number: 52479-85-3
MDL Number: MFCD00082966
Molecular Formula: C13H10O7
Molecular Weight: 278.22
Purity/Analysis Method: 98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 280
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR