(4-Hydroxyphenyl)diphenyl methanol ≥98.0% (by HPLC)
Supplier: TCI America
Synonyms:
4-Hydroxytrityl Alcohol
H1059-1G
TCH1059-1GEA
341
USD
TCH1059-1G
(4-Hydroxyphenyl)diphenyl methanol ≥98.0% (by HPLC)
(4-Hydroxyphenyl)diphenyl methanol
CAS Number: 15658-11-4
Molecular Formula: C19H16O2
Molecular Weight: 276.34
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 162
Storage Temperature: 0-10°C
Other Product Names / Synonyms: 4-Hydroxytrityl Alcohol
Formula:
C₁₉H₁₆O₂ MW: 276.34 g/mol Melting Pt: 162 °C |
CAS Number:
15658-11-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...