Methyl-2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate ≥98.0%

Supplier: TCI America

Synonyms: 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetic Acid Methyl Ester

H1049-100MG H1049-1G
TCH1049-100MGEA 58 USD
TCH1049-100MG TCH1049-1G
Methyl-2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate ≥98.0%
Methyl-2-[4-(hydroxymethyl)-2-nitrophenoxy]acetate

CAS Number: 308815-81-8
MDL Number: MFCD04117920
Molecular Formula: C10H11NO6
Molecular Weight: 241.20
Purity/Analysis Method: 98.0% (GC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Linkers (Combinatorial Chemistry)
Other Product Names / Synonyms: 2-[4-(Hydroxymethyl)-2-nitrophenoxy]acetic Acid Methyl Ester

Formula: C₁₀H₁₁NO₆
MW: 241.20 g/mol
MDL Number: MFCD04117920
CAS Number: 308815-81-8
https://us.vwr.com/stibo/web/std.lang.all/10/66/28971066.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR