1,1'-Azobis(N,N-dimethylformamide) ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N1,N1,N2,N2-Tetramethyldiazene-1,2-dicarboxamide
, Diamide
CAS Number: 10465-78-8
MDL Number: MFCD00008318
Molecular Formula: C6H12N4O2
Molecular Weight: 172.19
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 112
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Mitsunobu Reaction
Other Product Names / Synonyms: Diamide, N,N,N',N'-Tetramethylazodicarboxamide, TMAD
Formula:
C₆H₁₂N₄O₂ MW: 172.19 g/mol Melting Pt: 112 °C |
MDL Number:
MFCD00008318 CAS Number: 10465-78-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...