1-(2-Hydroxyethyl)-3,3-dimethylindolino-6'-nitrobenzopyrylospiran ≥93.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
2-(3',3'-Dimethyl-6-nitrospiro[chromene-2,2'-indolin]-1'-yl)ethanol
, 1'-(2-Hydroxyethyl)-3',3'-dimethyl-6-nitrospiro[1(2H)-benzopyran-2,2'-indoline]
H1042-1G
H1042-5G
TCH1042-1GEA
142.1
USD
TCH1042-1G
TCH1042-5G
1-(2-Hydroxyethyl)-3,3-dimethylindolino-6'-nitrobenzopyrylospiran ≥93.0% (by HPLC, titration analysis)
1-(2-Hydroxyethyl)-3,3-dimethylindolino-6'-nitrobenzopyrylospiran
CAS Number: 16111-07-2
MDL Number: MFCD00369031
Molecular Formula: C20H20N2O4
Molecular Weight: 352.39
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 165
Product Categories: Materials Chemistry, Photochromic Compounds
Other Product Names / Synonyms: 1'-(2-Hydroxyethyl)-3',3'-dimethyl-6-nitrospiro[1(2H)-benzopyran-2,2'-indoline]
Formula:
C₂₀H₂₀N₂O₄ MW: 352.39 g/mol Melting Pt: 165 °C |
MDL Number:
MFCD00369031 CAS Number: 16111-07-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...