Ethyl perfluorononanoate ≥98.0%

Supplier: TCI America

H1038-5G
TCH1038-5GEA 113 USD
TCH1038-5G
Ethyl perfluorononanoate ≥98.0%
Ethyl perfluorononanoate

CAS Number: 30377-52-7
MDL Number: MFCD00039245
Molecular Formula: C11H5F17O2
Molecular Weight: 492.13
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid


Product Categories: Synthetic Organic Chemistry, Fluorous Compounds, Fluorinated Building Blocks
Other Product Names / Synonyms: Heptadecafluorononanoic Acid Ethyl Ester, Ethyl Perfluorononanoate, Perfluorononanoic Acid Ethyl Ester

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR