(ɑ,ɑ,ɑ,2,3,5,6-Heptafluoro-p-tolyl)hydrazine ≥98.0%

Supplier: TCI America

Synonyms: 2,3,5,6-Tetrafluoro-4-hydrazinobenzotrifluoride(Perfluoro-p-tolyl)hydrazine

H1034-5G
TCH1034-5GEA 158 USD
TCH1034-5G
(ɑ,ɑ,ɑ,2,3,5,6-Heptafluoro-p-tolyl)hydrazine ≥98.0%
(alpha,alpha,alpha,2,3,5,6-Heptafluoro-p-tolyl)hydrazine

CAS Number: 1868-85-5
MDL Number: MFCD00012368
Molecular Formula: C7H3F7N2
Molecular Weight: 248.10
Purity/Analysis Method: 98.0% (GC)
Form: Crystal
Melting point (°C): 80


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: (Perfluoro-p-tolyl)hydrazine, 2,3,5,6-Tetrafluoro-4-hydrazinobenzotrifluoride

Formula: C₇H₃F₇N₂
MW: 248.10 g/mol
Melting Pt: 80 °C
MDL Number: MFCD00012368
CAS Number: 1868-85-5
https://us.vwr.com/stibo/web/std.lang.all/10/52/28971052.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR