(R,R)-(-)-2,5-Hexanediol ≥98.0%
Supplier: TCI America
Synonyms:
(2R,5R)-2,5-Hexanediol
, (2R,5R)-Hexane-2,5-diol
, (R,R)-(-)-2,5-Dihydroxy hexane
CAS Number: 17299-07-9
MDL Number: MFCD00142339
Molecular Formula: C6H14O2
Molecular Weight: 118.18
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 54
Flash Point (°C): 101
Specific rotation [a]20/D: -25 deg (C=1, EtOH)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (2R,5R)-2,5-Dihydroxyhexane
Formula:
C₆H₁₄O₂ MW: 118.18 g/mol Melting Pt: 54 °C Flash Pt: 101 °C |
MDL Number:
MFCD00142339 CAS Number: 17299-07-9 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...