3'-Hydroxyflavanone ≥98.0% (by HPLC)

Supplier: TCI America

H1025-1G
TCH1025-1GEA 132 USD
TCH1025-1G
3'-Hydroxyflavanone ≥98.0% (by HPLC)
3'-Hydroxyflavanone

CAS Number: 92496-65-6
MDL Number: MFCD00017738
Molecular Formula: C15H12O3
Molecular Weight: 240.26
Purity/Analysis Method: 98.0% (HPLC)
Form: Crystal

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR