2-Hydroxyisophthalaldehyde ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,6-Diformylphenol

H1018-5G H1018-1G
TCH1018-1GEA 351 USD
TCH1018-1G TCH1018-5G
2-Hydroxyisophthalaldehyde ≥98.0% (by GC, titration analysis)
2-Hydroxyisophthalaldehyde

CAS Number: 3328-69-6
MDL Number: MFCD00839420
Molecular Formula: C8H6O3
Molecular Weight: 150.13
Purity/Analysis Method: 98.0% (GC,T)
Form: Crystal
Melting point (°C): 124
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2,6-Diformylphenol

Formula: C₈H₆O₃
Melting Pt: 124 °C
MDL Number: MFCD00839420
CAS Number: 3328-69-6
https://us.vwr.com/stibo/web/std.lang.all/10/41/28971041.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR