2-Chloroethyl acetate ≥99.0%

Supplier: TCI America

A0027-25ML
TCA0027-025MLEA 153.22 USD
TCA0027-025ML
2-Chloroethyl acetate ≥99.0%
2-Chloroethyl acetate

CAS Number: 542-58-5
MDL Number: MFCD00018963
Molecular Formula: C4H7ClO2
Molecular Weight: 122.55
Purity/Analysis Method: 99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 145
Flash Point (°C): 54
Specific Gravity (20/20): 1.16


Other Product Names / Synonyms: Acetic Acid 2-Chloroethyl Ester

Formula: C₄H₇ClO₂
Boiling Pt: 145 °C
Density: 1.16 (20 °C)
Flash Pt: 54 °C
UN: 2929
ADR: 6.1,II
https://us.vwr.com/stibo/web/std.lang.all/74/65/28927465.jpg

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR