1,2,7-Heptanetriol ≥95.0%

Supplier: TCI America

H0957-5G
TCH0957-005GEA 881 USD
TCH0957-005G
1,2,7-Heptanetriol ≥95.0%
1,2,7-Heptanetriol

CAS Number: 37939-50-7
MDL Number: MFCD01861288
Molecular Formula: C7H16O3
Molecular Weight: 148.20
Purity/Analysis Method: 95.0% (GC)
Form: Crystal

Formula: C₇H₁₆O₃
MDL Number: MFCD01861288
CAS Number: 37939-50-7

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR