(S)-(-)-ɑ-Hydroxy-γ-butyrolactone ≥97.0%

Synonyms: (S)-(-)-Dihydro-3-hydroxy-2(3H)-furanone

H0951-100MG
TCH0951-100MGEA 61.37 USD
TCH0951-100MG
(S)-(-)-ɑ-Hydroxy-γ-butyrolactone ≥97.0%
(S)-(-)-alpha-Hydroxy-gamma-butyrolactone

CAS Number: 52079-23-9
MDL Number: MFCD00211245
Molecular Formula: C4H6O3
Molecular Weight: 102.09
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 1.30
Specific rotation [a]20/D: -72 deg (C=1, CHCl3)


Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (S)-(-)-Dihydro-3-hydroxy-2(3H)-furanone

Formula: C₄H₆O₃
MW: 102.09 g/mol
Density: 1.30 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00211245
CAS Number: 52079-23-9
https://us.vwr.com/stibo/web/std.lang.all/09/73/28970973.jpg

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR