6-Hepten-1-ol ≥96.0%

Supplier: TCI America

H0949-5G H0949-25G
TCH0949-005GEA 490.34 USD
TCH0949-005G TCH0949-025G
6-Hepten-1-ol ≥96.0%
6-Hepten-1-ol

CAS Number: 4117-10-6
MDL Number: MFCD00229167
Molecular Formula: C7H14O
Molecular Weight: 114.19
Purity/Analysis Method: 96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Specific Gravity (20/20): 0.85

Formula: C₇H₁₄O
Boiling Pt: 113 °C
Density: 0.85 (20 °C)
MDL Number: MFCD00229167
CAS Number: 4117-10-6
https://us.vwr.com/stibo/web/std.lang.all/09/70/28970970.jpg

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR