1,2-Heptanediol ≥98.0%

Supplier: TCI America

Synonyms: 1,2-Dihydroxyheptane

H0948-5G
TCH0948-005GEA 167 USD
TCH0948-005G
1,2-Heptanediol ≥98.0%
1,2-Heptanediol

CAS Number: 3710-31-4
MDL Number: MFCD01861287
Molecular Formula: C7H16O2
Molecular Weight: 132.20
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 130
Specific Gravity (20/20): 0.94


Other Product Names / Synonyms: 1,2-Dihydroxyheptane

Formula: C₇H₁₆O₂
MW: 132.20 g/mol
Boiling Pt: 130 °C
Density: 0.94 (20 °C)
MDL Number: MFCD01861287
CAS Number: 3710-31-4
https://us.vwr.com/stibo/web/std.lang.all/09/69/28970969.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR