(S)-(+)-2-Hydroxy-4-phthalimidobutyric acid
Supplier: TCI America
Synonyms:
(S)-(+)-a-Hydroxy-1,3-dioxo-2-isoindolinebutyric acid
H0933-5G
TCH0933-005GEA
101.45
USD
TCH0933-005G
(S)-(+)-2-Hydroxy-4-phthalimidobutyric acid
(S)-(+)-2-Hydroxy-4-phthalimidobutyric acid
CAS Number: 48172-10-7
MDL Number: MFCD00192213
Molecular Formula: C12H11NO5
Molecular Weight: 249.22
Form: Crystal
Color: White
Melting point (°C): 152
Specific rotation [a]20/D: 6 deg (C=1, MeOH)
Formula:
C₁₂H₁₁NO₅ MW: 249.22 g/mol Melting Pt: 152 °C |
MDL Number:
MFCD00192213 CAS Number: 48172-10-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...