4'-Hydroxyhexanophenone ≥99.0%
Supplier: TCI America
Synonyms:
4'-Hydroxycaprophenone
H0884-5G
H0884-25G
TCH0884-025GEA
231.25
USD
TCH0884-025G
TCH0884-005G
4'-Hydroxyhexanophenone ≥99.0%
4'-Hydroxyhexanophenone
CAS Number: 2589-72-2
MDL Number: MFCD00671540
Molecular Formula: C12H16O2
Molecular Weight: 192.26
Purity/Analysis Method:
Form: Crystal
Boiling point (°C): 167
Melting point (°C): 63
Other Product Names / Synonyms: Amyl 4-Hydroxyphenyl Ketone, 4'-Hydroxycaprophenone
Formula:
C₁₂H₁₆O₂ Boiling Pt: 167 °C Melting Pt: 63 °C |
MDL Number:
MFCD00671540 CAS Number: 2589-72-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...