1,7-Diacetoxyheptane ≥95.0%

Synonyms: Heptamethylene Diacetate1,7-Heptanediol Diacetate

H0850-10G
TCH0850-010GEA 127.5 USD
TCH0850-010G
1,7-Diacetoxyheptane ≥95.0%
1,7-Diacetoxyheptane

CAS Number: 52819-34-8
MDL Number: MFCD00191657
Molecular Formula: C11H20O4
Molecular Weight: 216.28
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 270
Specific Gravity (20/20): 1.00


Other Product Names / Synonyms: Heptamethylene Diacetate, 1,7-Heptanediol Diacetate

Formula: C₁₁H₂₀O₄
MW: 216.28 g/mol
Boiling Pt: 270 °C
Density: 1.00 (20 °C)
MDL Number: MFCD00191657
CAS Number: 52819-34-8
https://us.vwr.com/stibo/web/std.lang.all/08/75/28970875.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR