6-Heptenyl acetate ≥97.0%

Supplier: TCI America

H0849-5ML
TCH0849-005MLEA 274 USD
TCH0849-005ML
6-Heptenyl acetate ≥97.0%
6-Heptenyl acetate

CAS Number: 5048-30-6
MDL Number: MFCD00191656
Molecular Formula: C9H16O2
Molecular Weight: 156.23
Purity/Analysis Method: 97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 74
Specific Gravity (20/20): 0.89


Other Product Names / Synonyms: 7-Acetoxy-1-heptene, Acetic Acid 6-Heptenyl Ester

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR