N-(8-Hydroxyoctyl)phthalimide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 8-(N-Phthalimidyl)-1-octanol

H0834-1G
TCH0834-001GEA 275 USD
TCH0834-001G
N-(8-Hydroxyoctyl)phthalimide ≥98.0% (by titrimetric analysis)
N-(8-Hydroxyoctyl)phthalimide

MDL Number: MFCD00191517
Molecular Formula: C16H21NO3
Molecular Weight: 275.35
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 65


Other Product Names / Synonyms: 8-(N-Phthalimidyl)-1-octanol

Formula: C₁₆H₂₁NO₃
MW: 275.35 g/mol
Melting Pt: 65 °C
MDL Number: MFCD00191517
CAS Number: 105264-63-9
https://us.vwr.com/stibo/web/std.lang.all/08/60/28970860.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR