1,5-Hexadiene-3,4-diol ≥95.0% (by GC) stabilized

Supplier: TCI America
Danger

Synonyms: 3,4-Dihydroxy-1,5-hexadieneDivinylethylene Glycol1,2-Divinylglycol

Stabilization: Stabilised with hydroquinone (HQ)

H0807-10ML
TCH0807-010MLEA 219.18 USD
TCH0807-010ML
1,5-Hexadiene-3,4-diol ≥95.0% (by GC) stabilized
1,5-Hexadiene-3,4-diol

(stabilized with HQ)
Form: Clear Liquid
Boiling point (°C): 198
Flash Point (°C): 92
Specific Gravity (20/20): 1.02
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 3,4-Dihydroxy-1,5-hexadiene, Divinylethylene Glycol, 1,2-Divinylglycol

Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081129
CAS Number: 1069-23-4
EINECS: 213-955-5
UN: 2810
ADR: 6.1,III

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR