3-Hydroxypicolinamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 3-Hydroxypyridine-2-carboxamide

H0786-5G
TCH0786-005GEA 186 USD
TCH0786-005G
3-Hydroxypicolinamide ≥98.0% (by titrimetric analysis)
3-Hydroxypicolinamide

CAS Number: 933-90-4
MDL Number: MFCD00006302
Molecular Formula: C6H6N2O2
Molecular Weight: 138.13
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Melting point (°C): 199


Other Product Names / Synonyms: 3-Hydroxy-2-pyridinecarboxamide

Formula: C₆H₆N₂O₂
Melting Pt: 199 °C
MDL Number: MFCD00006302
CAS Number: 933-90-4
https://us.vwr.com/stibo/web/std.lang.all/08/17/28970817.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR