Heptadecanophenone ≥98.0%
Supplier: TCI America
H0780-25G
TCH0780-025GEA
315.53
USD
TCH0780-025G
Heptadecanophenone ≥98.0%
Heptadecanophenone
CAS Number: 128189-46-8
MDL Number: MFCD00191443
Molecular Formula: C23H38O
Molecular Weight: 330.56
Purity/Analysis Method:
Form: Crystal
Melting point (°C): 57
Formula:
C₂₃H₃₈O Melting Pt: 57 °C |
MDL Number:
MFCD00191443 CAS Number: 128189-46-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...