4-Heptylbenzonitrile ≥98.0%

Supplier: TCI America

H0778-5ML H0778-1ML
TCH0778-005MLEA 181 USD
TCH0778-005ML TCH0778-1ML
4-Heptylbenzonitrile ≥98.0%
4-Heptylbenzonitrile

CAS Number: 60484-67-5
MDL Number: MFCD00143348
Molecular Formula: C14H19N
Molecular Weight: 201.31
Purity/Analysis Method: 98.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 0.92


Product Categories: Materials Chemistry, Building Blocks for Liquid Crystals
Other Product Names / Synonyms: 1-(4-Cyanophenyl)heptane

Formula: C₁₄H₁₉N
Density: 0.92 (20 °C)
MDL Number: MFCD00143348
CAS Number: 60484-67-5
https://us.vwr.com/stibo/web/std.lang.all/08/10/28970810.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR