2-Hydroxymethyloxetane ≥95.0%

Supplier: TCI America

Synonyms: 2,4-Epoxybutanol2-OxetanemethanolOxetan-2-ylmethanol

H0773-1G
TCH0773-001GEA 282.17 USD
TCH0773-001G
2-Hydroxymethyloxetane ≥95.0%
2-Hydroxymethyloxetane

CAS Number: 61266-70-4
MDL Number: MFCD00142933
Molecular Formula: C4H8O2
Molecular Weight: 88.11
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 98
Specific Gravity (20/20): 1.08
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 2,4-Epoxybutanol, 2-Oxetanemethanol

Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 98 °C
Density: 1.08 (20 °C)
MDL Number: MFCD00142933
CAS Number: 61266-70-4
https://us.vwr.com/stibo/web/std.lang.all/08/05/28970805.jpg

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR