HEPPSO(β-hydroxy-4-(2-hydroxyethyl)piperazine-1-propanesulfonic acid) ≥98.0% (by titrimetric analysis)
Supplier: TCI America
|
Danger
|
Synonyms:
3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulfonic acid
, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulphonic acid
[Good's buffer component for biological research]
CAS Number: 68399-78-0
MDL Number: MFCD00038351
Molecular Formula: C9H20N2O5S
Molecular Weight: 268.33
Purity/Analysis Method:
Form: Crystal
Product Categories: Bioscience, Good's Buffering Agents
Other Product Names / Synonyms: HEPPSO, 3-[4-(2-Hydroxyethyl)-1-piperazinyl]-2-hydroxypropanesulfonic Acid
Formula:
C₉H₂₀N₂O₅S MW: 268.3305 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00038351 CAS Number: 68399-78-0 EINECS: 269-990-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...