N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline

Supplier: TCI America

Synonyms: 2-Cyano-N-(2-hydroxyethyl)-4-nitroaniline2-(2-Hydroxyethylamino)-5-nitrobenzonitrile

H0749-5G
TCH0749-005GEA 90 USD
TCH0749-005G
N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline
N-(2-Hydroxyethyl)-2-cyano-4-nitroaniline

CAS Number: 63989-40-2
MDL Number: MFCD00051934
Molecular Formula: C9H9N3O3
Molecular Weight: 207.19
Form: Crystal
Color: Pale Yellow
Melting point (°C): 163


Other Product Names / Synonyms: 2-Cyano-N-(2-hydroxyethyl)-4-nitroaniline, 2-(2-Hydroxyethylamino)-5-nitrobenzonitrile

Formula: C₉H₉N₃O₃
MW: 207.19 g/mol
Melting Pt: 163 °C
MDL Number: MFCD00051934
CAS Number: 63989-40-2
https://us.vwr.com/stibo/web/std.lang.all/07/83/28970783.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR