TCIAH0706

H0706-25G
TCH0706-025GEA 122.58 USD
TCH0706-025G
TCIAH0706
CAS Number: 19757-97-2
MDL Number: MFCD00043941
Molecular Formula: C4H8O4
Molecular Weight: 120.10
Purity/Analysis Method: 97.0% (T)
Form: Clear Liquid
Flash Point (°C): 67

Other Product Names / Synonyms: Glyoxylic Acid Methyl Ester Methyl Hemiacetal, 2-Hydroxy-2-methoxyacetic Acid Methyl Ester
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR