cis-3-Hexenyl-cis-3-hexenoate ≥95.0%

Supplier: TCI America

Synonyms: cis-3-Hexenoic Acid cis-3-Hexenyl Ester

H0677-5ML
TCH0677-005MLEA 43.38 USD
TCH0677-005ML
cis-3-Hexenyl-cis-3-hexenoate ≥95.0%
cis-3-Hexenyl-cis-3-hexenoate

CAS Number: 61444-38-0
MDL Number: MFCD00036652
Molecular Formula: C12H20O2
Molecular Weight: 196.29
Purity/Analysis Method: 95.0% (GC)
Form: Clear Liquid
Flash Point (°C): 104
Specific Gravity (20/20): 0.91


Other Product Names / Synonyms: cis-3-Hexenoic Acid cis-3-Hexenyl Ester

Formula: C₁₂H₂₀O₂
MW: 196.29 g/mol
Density: 0.91 (20 °C)
Flash Pt: 104 °C
MDL Number: MFCD00036652
CAS Number: 61444-38-0

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR